1-Bromo-4-isobutoxynaphthalene ≥97%

Supplier: AOBChem USA

Warning

64382-5G 64382-25G
77368-254EA 152.44 USD
77368-254 77380-482
1-Bromo-4-isobutoxynaphthalene ≥97%
1-Bromo-4-isobutoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₄H₁₅BrO
MW: 279.1723 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761747
CAS Number: 1379588-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR