1-(4-(Trifluoromethyl)phenyl)cyclohexanol ≥97%

Supplier: AOBChem USA

Warning

64349-5G 64349-25G
77368-224EA 61.8 USD
77368-224 77380-474
1-(4-(Trifluoromethyl)phenyl)cyclohexanol ≥97%
1-(4-(Trifluoromethyl)phenyl)cyclohexanol

Precaución: for Research Use Only

Formula: C₁₃H₁₅F₃O
MW: 244.2528 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18022791
CAS Number: 29480-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR