2,4-Difluoro-3-iodo-1,5-dimethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

63708-5G 63708-25G
77368-070EA 95.79 USD
77368-070 77380-328
2,4-Difluoro-3-iodo-1,5-dimethoxybenzene ≥97%
2,4-Difluoro-3-iodo-1,5-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₇F₂IO₂
Storage Temperature: Ambient
MDL Number: MFCD32706360
CAS Number: 1560796-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR