2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone ≥97%

Supplier: AOBChem USA

Warning

63298-5G 63298-25G
77368-002EA 90.64 USD
77368-002 77380-214
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone ≥97%
2,2,2-Trifluoro-3'-(trifluoromethyl)acetophenone

Precaución: for Research Use Only

Formula: C₉H₄F₆O
Storage Temperature: Ambient
MDL Number: MFCD00061275
CAS Number: 721-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR