2,4-Difluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

62791-5G 62791-25G
77367-906EA 166.86 USD
77367-906 77380-088
2,4-Difluoro-[1,1'-biphenyl]-3-carboxylic acid ≥97%
2,4-Difluoro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈F₂O₂
MW: 234.1982 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34761044
CAS Number: 2294046-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR