4,6-Dibromo-2-chloro-3-(trifluoromethyl)phenol ≥95%

Supplier: AOBChem USA

Warning

62672-5G 62672-25G
77367-872EA 166.86 USD
77367-872 77380-014
4,6-Dibromo-2-chloro-3-(trifluoromethyl)phenol ≥95%
4,6-Dibromo-2-chloro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₂Br₂ClF₃O
MW: 354.3463 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760968
CAS Number: 2807455-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR