6-Bromo-2-chloro-3-(trifluoromethyl)phenol ≥95%

Supplier: AOBChem USA

Warning

62671-5G 62671-25G
77367-870EA 166.86 USD
77367-870 77380-012
6-Bromo-2-chloro-3-(trifluoromethyl)phenol ≥95%
6-Bromo-2-chloro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₃BrClF₃O
MW: 275.4503 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760967
CAS Number: 2090960-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR