2-Chloro-1-isopropoxy-3-(trifluoromethyl)benzene ≥95%

Supplier: AOBChem USA

Warning

62669-5G 62669-25G
77367-868EA 161.71 USD
77367-868 77380-010
2-Chloro-1-isopropoxy-3-(trifluoromethyl)benzene ≥95%
2-Chloro-1-isopropoxy-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₀ClF₃O
MW: 238.6340 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23114182
CAS Number: 1369944-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR