1-(Benzyloxy)-2-chloro-3-(trifluoromethyl)benzene ≥97%

Supplier: AOBChem USA

Warning

62668-5G 62668-25G
77367-866EA 166.86 USD
77367-866 77380-008
1-(Benzyloxy)-2-chloro-3-(trifluoromethyl)benzene ≥97%
1-(Benzyloxy)-2-chloro-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₀ClF₃O
MW: 286.6768 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34760966
CAS Number: 2807464-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR