3-Bromo-5-chloro-[1,1'-biphenyl]-4-ol ≥97%

Supplier: AOBChem USA

Warning

61669-5G 61669-25G
77367-470EA 152.44 USD
77367-470 77379-646
3-Bromo-5-chloro-[1,1'-biphenyl]-4-ol ≥97%
3-Bromo-5-chloro-[1,1'-biphenyl]-4-ol

Precaución: for Research Use Only

Formula: C₁₂H₈BrClO
MW: 283.5483 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26895867
CAS Number: 4336-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR