(2-Fluoro-5-(trifluoromethyl)phenyl)(morpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

61519-5G 61519-25G
77367-420EA 166.86 USD
77367-420 77379-602
(2-Fluoro-5-(trifluoromethyl)phenyl)(morpholino)methanone ≥95%
(2-Fluoro-5-(trifluoromethyl)phenyl)(morpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₄NO₂
Storage Temperature: Ambient
MDL Number: MFCD24930815
CAS Number: 1526572-06-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR