(2-Fluoro-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

61518-5G 61518-25G
77367-418EA 161.71 USD
77367-418 77379-600
(2-Fluoro-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone ≥97%
(2-Fluoro-5-(trifluoromethyl)phenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₁F₄NOS
MW: 293.2804 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24930854
CAS Number: 1521546-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR