1,2,3-Tribromo-5-isopropylbenzene ≥97%

Supplier: AOBChem USA

Warning

61510-5G 61510-25G
77367-412EA 161.71 USD
77367-412 77379-594
1,2,3-Tribromo-5-isopropylbenzene ≥97%
1,2,3-Tribromo-5-isopropylbenzene

Precaución: for Research Use Only

Formula: C₉H₉Br₃
Storage Temperature: Ambient
MDL Number: MFCD34760383
CAS Number: 2764732-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR