3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-Bromo-4-fluoro-5-(trifluoromethyl)aniline

61404-5G 61404-25G
77367-390EA 74.16 USD
77367-390 77379-530
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine ≥97%
3-Bromo-4-fluoro-5-trifluoromethyl-phenylamine

Precaución: for Research Use Only

Formula: C₇H₄BrF₄N
MW: 258.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11520114
CAS Number: 1233026-11-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR