2,3,4-Trifluoro-1,5-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

61145-5G 61145-25G
77367-332EA 100.94 USD
77367-332 77379-434
2,3,4-Trifluoro-1,5-diiodobenzene ≥97%
2,3,4-Trifluoro-1,5-diiodobenzene

Precaución: for Research Use Only

Formula: C₆HF₃I₂
MW: 383.8762 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178320
CAS Number: 459425-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR