1-(2-Chloroisonicotinoyl)-4-methylpiperazine hydrochloride ≥97%

Supplier: AOBChem USA

Warning

60964-5G 60964-25G
77367-274EA 154.5 USD
77367-274 77379-378
1-(2-Chloroisonicotinoyl)-4-methylpiperazine hydrochloride ≥97%
1-(2-Chloroisonicotinoyl)-4-methylpiperazine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₅Cl₂N₃O
Storage Temperature: Ambient
MDL Number: MFCD09864559
CAS Number: 612487-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR