2-Chloro-N-cyclopropylisonicotinamide ≥97%

Supplier: AOBChem USA

Warning

60958-5G 60958-25G
77367-266EA 154.5 USD
77367-266 77379-370
2-Chloro-N-cyclopropylisonicotinamide ≥97%
2-Chloro-N-cyclopropylisonicotinamide

Precaución: for Research Use Only

Formula: C₉H₉ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD09864587
CAS Number: 959241-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR