4-Bromo-2-fluoro-1-((4-methoxybenzyl)oxy)benzene ≥97%

Supplier: AOBChem USA

Warning

60695-5G 60695-25G
77367-194EA 166.86 USD
77367-194 77379-288
4-Bromo-2-fluoro-1-((4-methoxybenzyl)oxy)benzene ≥97%
4-Bromo-2-fluoro-1-((4-methoxybenzyl)oxy)benzene

Precaución: for Research Use Only

Formula: C₁₄H₁₂BrFO₂
MW: 311.146 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759891
CAS Number: 2755724-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR