1,3-Dibromonaphthalene ≥95%

Supplier: AOBChem USA

Warning

60631-5G 60631-25G
77367-162EA 62.83 USD
77367-162 77379-272
1,3-Dibromonaphthalene ≥95%
1,3-Dibromonaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆Br₂
Storage Temperature: Ambient
MDL Number: MFCD18411509
CAS Number: 52358-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR