4-(Benzyloxy)-2-fluoro-1-methoxybenzene ≥95%

Supplier: AOBChem USA

Warning

60501-5G 60501-25G
77367-108EA 181.28 USD
77367-108 77379-208
4-(Benzyloxy)-2-fluoro-1-methoxybenzene ≥95%
4-(Benzyloxy)-2-fluoro-1-methoxybenzene

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759780
CAS Number: 1709852-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR