(3,5-Dimethoxyphenyl)(piperidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

60448-5G 60448-25G
77367-092EA 166.86 USD
77367-092 77379-182
(3,5-Dimethoxyphenyl)(piperidin-1-yl)methanone ≥97%
(3,5-Dimethoxyphenyl)(piperidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₃
MW: 249.306 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00751711
CAS Number: 349134-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR