2-(2,2-Dichloro-1-hydroxyethyl)-4-methylphenol ≥95%

Supplier: AOBChem USA

Warning

60414-5G 60414-25G
77367-080EA 166.86 USD
77367-080 77379-176
2-(2,2-Dichloro-1-hydroxyethyl)-4-methylphenol ≥95%
2-(2,2-Dichloro-1-hydroxyethyl)-4-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₀Cl₂O₂
MW: 221.081 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759740
CAS Number: 2807454-94-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR