2-Bromo-5-chlorophenyl acetate ≥97%

Supplier: AOBChem USA

Warning

60353-5G 60353-25G
77367-068EA 181.28 USD
77367-068 77379-152
2-Bromo-5-chlorophenyl acetate ≥97%
2-Bromo-5-chlorophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.489 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34759703
CAS Number: 142148-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR