1-Fluoro-2-isopropoxy-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

59820-5G 59820-25G
77366-916EA 166.86 USD
77366-916 77378-964
1-Fluoro-2-isopropoxy-4-methylbenzene ≥97%
1-Fluoro-2-isopropoxy-4-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20429968
CAS Number: 1344249-63-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR