2,4-Dibromo-6-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

59321-5G 59321-25G
77366-728EA 106.09 USD
77366-728 77378-790
2,4-Dibromo-6-(trifluoromethoxy)phenol ≥97%
2,4-Dibromo-6-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₃Br₂F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD25954478
CAS Number: 1027269-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR