(E)-4-Chloro-2-fluorobenzaldehyde oxime ≥97%

Supplier: AOBChem USA

Warning

58870-5G 58870-25G
77366-638EA 87.55 USD
77366-638 77378-624
(E)-4-Chloro-2-fluorobenzaldehyde oxime ≥97%
(E)-4-Chloro-2-fluorobenzaldehyde oxime

Precaución: for Research Use Only

Formula: C₇H₅ClFNO
Storage Temperature: Ambient
MDL Number: MFCD00143283
CAS Number: 1629614-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR