1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

57240-5G 57240-25G
77366-190EA 132.87 USD
77366-190 77378-082
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid ≥97%
1,3,5-Trimethyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00159648
CAS Number: 1125-29-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR