(2-Fluoro-5-methylphenyl)(thiomorpholino)methanone ≥97%

Supplier: AOBChem USA

Warning

56979-5G 56979-25G
77366-116EA 152.44 USD
77366-116 77378-004
(2-Fluoro-5-methylphenyl)(thiomorpholino)methanone ≥97%
(2-Fluoro-5-methylphenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄FNOS
MW: 239.309 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16783515
CAS Number: 1249275-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR