N-Cyclohexyl-2-fluoro-5-methylbenzamide ≥97%

Supplier: AOBChem USA

Warning

56975-5G 56975-25G
77366-110EA 173.04 USD
77366-110 77378-000
N-Cyclohexyl-2-fluoro-5-methylbenzamide ≥97%
N-Cyclohexyl-2-fluoro-5-methylbenzamide

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO
Storage Temperature: Ambient
MDL Number: MFCD17498385
CAS Number: 1311624-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR