2-Bromo-5-fluorophenyl acetate ≥95%

Supplier: AOBChem USA

Warning

56547-5G 56547-25G
77365-974EA 181.28 USD
77365-974 77377-874
2-Bromo-5-fluorophenyl acetate ≥95%
2-Bromo-5-fluorophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD34530531
CAS Number: 1456627-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR