tert-Butyl 3,4-difluoro-2-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

56316-5G 56316-25G
77365-912EA 166.86 USD
77365-912 77377-806
tert-Butyl 3,4-difluoro-2-methylbenzoate ≥97%
tert-Butyl 3,4-difluoro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O₂
MW: 228.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757775
CAS Number: 2755719-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR