1-(3-Bromo-2-methylphenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

55584-5G 55584-25G
77365-792EA 145.23 USD
77365-792 77377-638
1-(3-Bromo-2-methylphenyl)ethanone ≥97%
1-(3-Bromo-2-methylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22573012
CAS Number: 52779-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR