Ethyl 3-hydroxydecanoate ≥97%

Supplier: AOBChem USA

Warning

55394-5G 55394-25G
77365-718EA 135.96 USD
77365-718 77377-566
Ethyl 3-hydroxydecanoate ≥97%
Ethyl 3-hydroxydecanoate

Precaución: for Research Use Only

Formula: C₁₂H₂₄O₃
MW: 216.317 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066353
CAS Number: 6071-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR