4-(1-Hydroxypropyl)benzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 1-[4-(Trifluoromethyl)phenyl]propanol, 1-(4-(Trifluoromethyl)phenyl)propan-1-ol

54949-5G 54949-25G
77365-662EA 133.9 USD
77365-662 77377-490
4-(1-Hydroxypropyl)benzotrifluoride ≥97%
4-(1-Hydroxypropyl)benzotrifluoride

Precaución: for Research Use Only

Formula: C₁₀H₁₁F₃O
MW: 204.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07773046
CAS Number: 67081-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR