2-Ethyl-4-iodo-6-methylphenol ≥97%

Supplier: AOBChem USA

Warning

54820-5G 54820-25G
77365-634EA 132.87 USD
77365-634 77377-460
2-Ethyl-4-iodo-6-methylphenol ≥97%
2-Ethyl-4-iodo-6-methylphenol

Precaución: for Research Use Only

Formula: C₉H₁₁IO
Storage Temperature: Ambient
MDL Number: MFCD34757391
CAS Number: 2710967-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR