2-(Benzyloxy)-5-iodo-1,3-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

54745-5G 54745-25G
77365-616EA 131.84 USD
77365-616 77377-424
2-(Benzyloxy)-5-iodo-1,3-dimethylbenzene ≥95%
2-(Benzyloxy)-5-iodo-1,3-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅IO
Storage Temperature: Ambient
MDL Number: MFCD18072891
CAS Number: 877603-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR