9-Phenyl-9H-carbazole-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

54618-5G 54618-25G
77365-592EA 70.04 USD
77365-592 77377-386
9-Phenyl-9H-carbazole-3-carboxylic acid ≥95%
9-Phenyl-9H-carbazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₃NO₂
MW: 287.312 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530288
CAS Number: 2195344-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR