(4-Methyl-3-nitrophenyl)(thiomorpholino)methanone ≥95%

Supplier: AOBChem USA

Warning

54614-5G 54614-25G
77365-588EA 131.84 USD
77365-588 77377-382
(4-Methyl-3-nitrophenyl)(thiomorpholino)methanone ≥95%
(4-Methyl-3-nitrophenyl)(thiomorpholino)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃S
MW: 266.316 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13532439
CAS Number: 1197533-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR