(4-Methyl-3-nitrophenyl)(pyrrolidin-1-yl)methanone ≥97%

Supplier: AOBChem USA

Warning

54610-5G 54610-25G
77365-580EA 131.84 USD
77365-580 77377-376
(4-Methyl-3-nitrophenyl)(pyrrolidin-1-yl)methanone ≥97%
(4-Methyl-3-nitrophenyl)(pyrrolidin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂O₃
MW: 234.251 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00783664
CAS Number: 329059-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR