Methyl-1-benzofuran-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

54395-5G 54395-25G
77365-526EA 139.05 USD
77365-526 77377-296
Methyl-1-benzofuran-2-carboxylate ≥95%
Methyl-1-benzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD01159618
CAS Number: 1646-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR