4-Chloro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

54102-5G 54102-25G
77365-466EA 105.06 USD
77365-466 77377-240
4-Chloro-3-hydroxybenzaldehyde ≥97%
4-Chloro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD16999770
CAS Number: 56962-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR