3-Chloro-N-cyclopropyl-2,4-difluorobenzamide ≥97%

Supplier: AOBChem USA

Warning

53960-5G 53960-25G
77365-438EA 90.64 USD
77365-438 77377-212
3-Chloro-N-cyclopropyl-2,4-difluorobenzamide ≥97%
3-Chloro-N-cyclopropyl-2,4-difluorobenzamide

Precaución: for Research Use Only

Formula: C₁₀H₈ClF₂NO
MW: 231.626 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757187
CAS Number: 2764733-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR