3-Chloro-2,4-difluoro-N,N-dimethylbenzamide ≥97%

Supplier: AOBChem USA

Warning

53958-5G 53958-25G
77365-434EA 90.64 USD
77365-434 77377-208
3-Chloro-2,4-difluoro-N,N-dimethylbenzamide ≥97%
3-Chloro-2,4-difluoro-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₈ClF₂NO
MW: 219.616 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34757185
CAS Number: 2764730-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR