(2-Fluoro-6-iodo-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

53835-5G 53835-25G
77365-416EA 131.84 USD
77365-416 77377-144
(2-Fluoro-6-iodo-3-methylphenyl)(methyl)sulfane ≥97%
(2-Fluoro-6-iodo-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIS
MW: 282.117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29040760
CAS Number: 1935558-18-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR