4-(4-Bromophenoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

53719-5G 53719-25G
77365-392EA 183.34 USD
77365-392 77377-126
4-(4-Bromophenoxy)benzoic acid ≥97%
4-(4-Bromophenoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₃
MW: 293.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01244945
CAS Number: 21120-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR