1-Bromo-5-chloro-3-fluoro-2-isopropoxybenzene ≥97%

Supplier: AOBChem USA

Warning

53304-5G 53304-25G
77365-256EA 166.86 USD
77365-256 77376-990
1-Bromo-5-chloro-3-fluoro-2-isopropoxybenzene ≥97%
1-Bromo-5-chloro-3-fluoro-2-isopropoxybenzene

Precaución: for Research Use Only

Formula: C₉H₉BrClFO
MW: 267.523 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530210
CAS Number: 2734772-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR