2-(6-Bromopyridin-2-yl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

53171-5G 53171-25G
77365-230EA 133.9 USD
77365-230 77376-902
2-(6-Bromopyridin-2-yl)propan-2-ol ≥95%
2-(6-Bromopyridin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₈H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD16657880
CAS Number: 638218-78-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR