(4-Chloro-2-methoxyphenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

53120-5G 53120-25G
77365-222EA 161.71 USD
77365-222 77376-880
(4-Chloro-2-methoxyphenyl)(methyl)sulfane ≥95%
(4-Chloro-2-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₉ClOS
MW: 188.674 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178669
CAS Number: 23385-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR