3,5-Dibromo-[1,1'-biphenyl]-2-ol ≥97%

Supplier: AOBChem USA

Warning

53071-5G 53071-25G
77365-196EA 80.34 USD
77365-196 77376-862
3,5-Dibromo-[1,1'-biphenyl]-2-ol ≥97%
3,5-Dibromo-[1,1'-biphenyl]-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₈Br₂O
Storage Temperature: Ambient
MDL Number: MFCD00009714
CAS Number: 55815-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR