1-Bromo-5-iodo-2,4-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

52937-5G 52937-25G
77365-160EA 183.34 USD
77365-160 77376-806
1-Bromo-5-iodo-2,4-dimethylbenzene ≥97%
1-Bromo-5-iodo-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₈H₈BrI
MW: 310.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28738489
CAS Number: 854861-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR